Assay Details
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839770.1 | XP_028695603.1 | 1048 | 71 | ||
XM_028839771.1 | XP_028695604.1 | 1048 | 71 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839770.1 | XP_028695603.1 | 1048 | 71 | ||
XM_028839771.1 | XP_028695604.1 | 1048 | 71 |