Assay Details
Gene Symbol: ALDH3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833153.1 | XP_028688986.1 | 1212 | 92 | ||
XM_028833154.1 | XP_028688987.1 | 896 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
