Assay Details
Gene Symbol: LOC713438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140965.2 | XP_014996451.2 | 94 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015140965.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
