Assay Details
Gene Symbol: LOC106992664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110547.2 | XP_014966033.2 | 519 | 103 |
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110498.2 | XP_014965984.1 | 279 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015110498.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
