Assay Details
Gene Symbol: APOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090079.4 | XP_001090079.2 | 113 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001090079.4 transcripts.
Set Membership: |
![]() ![]() |
