Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836402.1 | XP_028692235.1 | 2658 | 63 | ||
XM_028836403.1 | XP_028692236.1 | 1115 | 63 | |||
XM_028836404.1 | XP_028692237.1 | 1179 | 63 | |||
XM_028836405.1 | XP_028692238.1 | 637 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
