Assay Details
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022522.2 | NP_071967.2 | 10 - 11 | 1236 | 59 | |
XM_006236394.2 | XP_006236456.1 | 9 - 10 | 1917 | 59 | ||
GenBank mRNA | AF136231.1 | - | 10 - 11 | 1234 | 59 | |
AF136232.1 | - | 11 - 12 | 1295 | 59 | ||
U77933.1 | - | 10 - 11 | 1236 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
luteolysis
neural retina development
proteolysis
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
brain development
aging
intrinsic apoptotic signaling pathway in response to DNA damage
programmed cell death
protein processing
ectopic germ cell programmed cell death
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
cellular response to mechanical stimulus
apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of apoptotic signaling pathway
molecular_function
cysteine-type endopeptidase activity
protein binding
protein domain specific binding
cysteine-type endopeptidase activity involved in apoptotic process
luteolysis
neural retina development
proteolysis
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
brain development
aging
intrinsic apoptotic signaling pathway in response to DNA damage
programmed cell death
protein processing
ectopic germ cell programmed cell death
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
cellular response to mechanical stimulus
apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of apoptotic signaling pathway
molecular_function
cysteine-type endopeptidase activity
protein binding
protein domain specific binding
cysteine-type endopeptidase activity involved in apoptotic process
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164041 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1019923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1019923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164041 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158644 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1019923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158644 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164041 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158644 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164041 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1019923 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1019923 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700144907 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30096 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73280 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700144907 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10855026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855024 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855022 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855023 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855021 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855025 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855027 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10855020 | - | - |
Rat Genome 230 2.0 Array | 1367891_a_at | - | - |
Rat Genome 230 2.0 Array | 1367891_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1367890_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367890_at | - | - |
Affymetrix HT Rat Focus Array | 1367891_a_at | - | - |
Affymetrix HT Rat Focus Array | 1367890_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367890_at | - | - |
Affymetrix HT Rat Focus Array | 1367891_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400005542.rn.1 | TC0400000752.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400005541.rn.1 | TC0400000752.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400005543.rn.1 | TC0400000752.rn.1 | RTA TC |
Rat Expression Set 230 | 1367890_PM_at | - | - |
Rat Expression Set 230 | 1367890_at | - | - |
Rat Expression Set 230 | 1367891_PM_a_at | - | - |
Rat Expression Set 230 | 1367891_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855020 | - | - |
Rat Genome U34 Set | U77933_at | - | - |
Rat Genome U34 Set | U34684_s_at | - | - |
Rat Genome U34 Set | AF025671_s_at | - | - |
Rat Genome U34 Set | AF025671_s_at | - | - |
Rat Genome U34 Set | U34684_s_at | - | - |
Rat Genome U34 Set | U77933_at | - | - |
Rat Genome U34 Set | U34684_s_at | - | - |
Rat Genome U34 Set | U77933_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22061218 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349906 | - | - |
RatRef-12 Expression BeadChip | ILMN_66477 | - | - |
