Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030023.1 | NP_001025194.1 | 7 - 8 | 880 | 107 | |
GenBank mRNA | AY880882.1 | - | 7 - 8 | 880 | 107 | |
KC795688.1 | - | 8 - 9 | 913 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
