Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 5 - 6 | 968 | 81 | |
XM_006256604.2 | XP_006256666.1 | 6 - 7 | 1256 | 81 | ||
XM_008772994.1 | XP_008771216.1 | 6 - 7 | 1096 | 81 | ||
XM_008772995.1 | XP_008771217.1 | 6 - 7 | 2445 | 81 | ||
XM_008772996.1 | XP_008771218.1 | 6 - 7 | 1079 | 81 | ||
XM_008772997.1 | XP_008771219.1 | 5 - 6 | 4721 | 81 | ||
XM_008772998.1 | XP_008771220.1 | 3 - 4 | 253 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
