Assay Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019328.3 | NP_062201.2 | 6 - 7 | 1654 | 71 | |
XM_006234201.2 | XP_006234263.1 | 7 - 8 | 1872 | 71 | ||
XM_008761861.1 | XP_008760083.1 | 6 - 7 | 1788 | 71 | ||
GenBank mRNA | DQ092629.1 | - | 7 - 8 | 1481 | 71 | |
DQ092630.1 | - | 7 - 8 | 1317 | 71 | ||
DQ092631.1 | - | 6 - 7 | 1478 | 71 | ||
DQ092632.1 | - | 6 - 7 | 1535 | 71 | ||
DQ092633.1 | - | 6 - 7 | 1421 | 71 | ||
FQ222109.1 | - | 7 - 8 | 1773 | 71 | ||
L08595.1 | - | 6 - 7 | 1651 | 71 | ||
U01146.1 | - | 6 - 7 | 1585 | 71 | ||
U72345.1 | - | 7 - 8 | 1878 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
