Assay Details
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 9 - 10 | 1727 | 70 | |
XM_006241020.2 | XP_006241082.1 | 10 - 11 | 1886 | 70 | ||
GenBank mRNA | AF029109.1 | - | 9 - 10 | 1727 | 70 | |
BC161842.1 | - | 10 - 11 | 1789 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
