Assay Details
Gene Symbol: Psmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017284.1 | NP_058980.1 | 1 - 2 | 154 | 90 | |
GenBank mRNA | BC058487.1 | - | 1 - 2 | 174 | 90 | |
D21799.1 | - | 1 - 2 | 154 | 90 | ||
FQ217737.1 | - | 1 - 2 | 184 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
