Assay Details
Gene Symbol: Npl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013984.1 | NP_001014006.1 | 2 - 3 | 141 | 66 | |
XM_006249997.2 | XP_006250059.1 | 2 - 3 | 289 | 66 | ||
GenBank mRNA | BC082004.1 | - | 2 - 3 | 141 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
