Assay Details
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057122.1 | NP_476463.1 | 4 - 5 | 485 | 90 | |
GenBank mRNA | BC063145.1 | - | 4 - 5 | 495 | 90 | |
D50695.1 | - | 4 - 5 | 485 | 90 | ||
FQ225557.1 | - | 4 - 5 | 510 | 90 | ||
FQ226113.1 | - | 4 - 5 | 494 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
