Assay Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033897.1 | NP_001029069.1 | 15 - 16 | 2021 | 78 | |
XM_006239584.2 | XP_006239646.1 | 16 - 17 | 1935 | 78 | ||
GenBank mRNA | BC101911.1 | - | 15 - 16 | 2021 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
