Assay Details
Gene Symbol: Parvb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134780.1 | NP_001128252.1 | 11 - 12 | 983 | 64 | |
XM_006241404.2 | XP_006241466.1 | 11 - 12 | 1056 | 64 | ||
XM_006241405.2 | XP_006241467.1 | 11 - 12 | 925 | 64 | ||
XM_006241406.2 | XP_006241468.1 | 11 - 12 | 2705 | 64 | ||
XM_006241407.2 | XP_006241469.1 | 11 - 12 | 1161 | 64 | ||
XM_006241409.2 | XP_006241471.1 | 11 - 12 | 899 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
