Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270701.1 | NP_001257630.1 | 2 - 3 | 394 | 75 | |
NM_001270703.1 | NP_001257632.1 | 2 - 3 | 394 | 75 | ||
NM_023021.2 | NP_075410.2 | 2 - 3 | 394 | 75 | ||
XM_006228466.2 | XP_006228528.1 | 2 - 3 | 2154 | 75 | ||
XM_006228467.2 | XP_006228529.1 | 2 - 3 | 2155 | 75 | ||
XM_006228469.1 | XP_006228531.1 | 3 - 4 | 376 | 75 | ||
XM_006228470.2 | XP_006228532.1 | 2 - 3 | 224 | 75 | ||
XM_006228471.1 | XP_006228533.1 | 2 - 3 | 2651 | 75 | ||
XM_008758975.1 | XP_008757197.1 | 3 - 4 | 2049 | 75 | ||
XM_008758976.1 | XP_008757198.1 | 2 - 3 | 573 | 75 | ||
XM_008758978.1 | XP_008757200.1 | 2 - 3 | 213 | 75 | ||
GenBank mRNA | AB292803.1 | - | 2 - 3 | 252 | 75 | |
AF149250.1 | - | 2 - 3 | 252 | 75 | ||
AF156554.1 | - | 2 - 3 | 394 | 75 | ||
AF190458.1 | - | 2 - 3 | 364 | 75 | ||
AJ133438.2 | - | 2 - 3 | 252 | 75 | ||
BC128736.1 | - | 2 - 3 | 345 | 75 | ||
EU872449.1 | - | 2 - 3 | 367 | 75 | ||
EU872450.1 | - | 2 - 3 | 367 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
