Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004068.1 | NP_001004068.1 | 1 - 2 | 302 | 73 | |
XM_006246973.2 | XP_006247035.1 | 1 - 2 | 610 | 73 | ||
XM_006246978.2 | XP_006247040.1 | 1 - 2 | 610 | 73 | ||
GenBank mRNA | BC079216.1 | - | 1 - 2 | 302 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079216.1, NM_001004068.1, XM_006246973.2, XM_006246978.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
