Assay Details
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013089.2 | NP_001013107.1 | 3 - 4 | 399 | 52 | |
XM_006238033.2 | XP_006238095.1 | 3 - 4 | 323 | 52 | ||
XM_008763615.1 | XP_008761837.1 | 2 - 3 | 168 | 52 | ||
GenBank mRNA | BC098756.1 | - | 3 - 4 | 399 | 52 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008763615.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
