Assay Details
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011929.1 | NP_001011929.1 | 5 - 6 | 886 | 116 | |
XM_006233435.2 | XP_006233497.1 | 5 - 6 | 898 | 116 | ||
GenBank mRNA | BC079085.1 | - | 5 - 6 | 886 | 116 | |
FQ216102.1 | - | 1 - 2 | 100 | 116 | ||
FQ234013.1 | - | 5 - 6 | 609 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
