Assay Details
Gene Symbol: Dcaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108716.1 | NP_001102186.1 | 10 - 11 | 1032 | 87 | |
XM_006240347.2 | XP_006240409.1 | 10 - 11 | 1035 | 87 | ||
XM_008764814.1 | XP_008763036.1 | 9 - 10 | 1083 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
