Assay Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004280.1 | NP_001004280.1 | 2 - 3 | 113 | 60 | |
XM_006234158.2 | XP_006234220.1 | 2 - 3 | 232 | 60 | ||
GenBank mRNA | BC078953.1 | - | 2 - 3 | 113 | 60 | |
FQ212704.1 | - | 2 - 3 | 108 | 60 | ||
FQ213711.1 | - | 2 - 3 | 90 | 60 | ||
FQ216239.1 | - | 2 - 3 | 108 | 60 | ||
FQ219117.1 | - | 2 - 3 | 109 | 60 | ||
FQ228704.1 | - | 2 - 3 | 109 | 60 | ||
FQ229971.1 | - | 2 - 3 | 78 | 60 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078953.1, NM_001004280.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
