Assay Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109445.1 | NP_001102915.1 | 2 - 3 | 193 | 79 | |
XM_006240399.2 | XP_006240461.1 | 2 - 3 | 167 | 79 | ||
GenBank mRNA | BC158833.1 | - | 2 - 3 | 135 | 79 | |
FJ179410.1 | - | 1 - 2 | 70 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
