Assay Details
Gene Symbol: Cnot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107313.1 | NP_001100783.1 | 2 - 3 | 412 | 98 | |
XM_008771290.1 | XP_008769512.1 | 2 - 3 | 585 | 98 | ||
XM_008771291.1 | XP_008769513.1 | 2 - 3 | 555 | 98 | ||
XM_008771292.1 | XP_008769514.1 | 2 - 3 | 455 | 98 | ||
GenBank mRNA | BC167766.1 | - | 2 - 3 | 371 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
