Assay Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 1 - 2 | 123 | 89 | |
GenBank mRNA | FM116382.1 | - | 1 - 2 | 82 | 89 | |
FM118664.1 | - | 1 - 2 | 107 | 89 | ||
FQ215476.1 | - | 1 - 2 | 118 | 89 | ||
FQ216044.1 | - | 1 - 2 | 122 | 89 | ||
FQ216100.1 | - | 1 - 2 | 99 | 89 | ||
FQ216185.1 | - | 1 - 2 | 120 | 89 | ||
FQ224925.1 | - | 1 - 2 | 120 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
