Assay Details
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134468.1 | NP_604463.1 | 6 - 7 | 673 | 89 | |
XM_006236999.2 | XP_006237061.1 | 7 - 8 | 1342 | 89 | ||
XM_006237000.2 | XP_006237062.1 | 7 - 8 | 1338 | 89 | ||
XM_006237001.2 | XP_006237063.1 | 6 - 7 | 1205 | 89 | ||
XM_006237002.2 | XP_006237064.1 | 6 - 7 | 1132 | 89 | ||
XM_006237003.2 | XP_006237065.1 | 6 - 7 | 1131 | 89 | ||
GenBank mRNA | BC071177.1 | - | 6 - 7 | 608 | 89 | |
FQ213485.1 | - | 636 | 89 | |||
L24907.1 | - | 6 - 7 | 647 | 89 | ||
L26288.1 | - | 6 - 7 | 673 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
