Assay Details
Gene Symbol: Lpxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009649.1 | NP_001009649.1 | 6 - 7 | 732 | 66 | |
GenBank mRNA | BC088217.1 | - | 6 - 7 | 732 | 66 | |
FQ230386.1 | - | 6 - 7 | 697 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
