Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127544.1 | NP_001121016.1 | 4 - 5 | 575 | 127 | |
XM_008761404.1 | XP_008759626.1 | 3 - 4 | 603 | 127 | ||
GenBank mRNA | BC162022.1 | - | 4 - 5 | 554 | 127 | |
FQ211892.1 | - | 3 - 4 | 535 | 127 | ||
FQ221591.1 | - | 4 - 5 | 655 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
