Assay Details
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012568.2 | NP_036700.1 | 6 - 7 | 1265 | 74 | |
XM_003752039.3 | XP_003752087.1 | 7 - 8 | 889 | 74 | ||
XM_008773152.1 | XP_008771374.1 | 6 - 7 | 1258 | 74 | ||
XM_008773153.1 | XP_008771375.1 | 6 - 7 | 671 | 74 | ||
GenBank mRNA | AJ310837.1 | - | 6 - 7 | 714 | 74 | |
X61159.1 | - | 6 - 7 | 1265 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10629 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053636 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053636 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053636 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P395915 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82975 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49149 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28962 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103450 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41978 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62120 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13509 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53721 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31540 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77015 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40250 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48577 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50314 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23185 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59980 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76850 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22047 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10933394 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933396 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933400 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933397 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933399 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933404 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933395 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933401 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10933393 | - | - |
Rat Genome 230 2.0 Array | 1371153_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1387696_a_at | - | - |
Rat Genome 230 2.0 Array | 1387696_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1371153_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371153_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387696_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371153_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387696_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00010487.rn.1 | TC0X00001624.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00010491.rn.1 | TC0X00001624.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00010490.rn.1 | TC0X00001624.rn.1 | RTA TC |
Rat Expression Set 230 | 1371153_a_at | - | - |
Rat Expression Set 230 | 1387696_a_at | - | - |
Rat Expression Set 230 | 1371153_PM_a_at | - | - |
Rat Expression Set 230 | 1387696_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933393 | - | - |
Rat Genome U34 Set | X57281_at | - | - |
Rat Genome U34 Set | X57281_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22196670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376728 | - | - |
