Assay Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 1 - 2 | 243 | 95 | ||
XM_006239907.2 | XP_006239969.1 | 1 - 2 | 246 | 95 | ||
GenBank mRNA | M16982.1 | - | 1 - 2 | 301 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M16982.1, XM_006239906.2, XM_006239907.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
