Assay Details
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012637.2 | NP_036769.1 | 3 - 4 | 336 | 56 | |
XM_006235638.2 | XP_006235700.1 | 3 - 4 | 405 | 56 | ||
XM_006235639.2 | XP_006235701.1 | 3 - 4 | 406 | 56 | ||
GenBank mRNA | BC081829.1 | - | 3 - 4 | 336 | 56 | |
M33962.1 | - | 3 - 4 | 378 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
