Assay Details
Gene Symbol: Scn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012647.1 | NP_036779.1 | 11 - 12 | 1878 | 79 | |
GenBank mRNA | M22254.1 | - | 11 - 12 | 1878 | 79 | |
X03639.1 | - | 11 - 12 | 1878 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
