Assay Details
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012727.3 | NP_036859.2 | 12 - 13 | 1199 | 102 | |
XM_006254523.2 | XP_006254585.1 | 11 - 12 | 1141 | 102 | ||
GenBank mRNA | BC128706.1 | - | 12 - 13 | 1199 | 102 | |
J04446.1 | - | 2 - 3 | 274 | 102 | ||
J04600.1 | - | 226 | 102 | |||
M63334.1 | - | 1093 | 102 | |||
M64757.1 | - | 10 - 11 | 1189 | 102 | ||
S65840.1 | - | 11 - 12 | 1124 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the J04446.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
myeloid dendritic cell cytokine production
protein phosphorylation
nucleocytoplasmic transport
inflammatory response
signal transduction
neuron-neuron synaptic transmission
long-term memory
myeloid dendritic cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
myeloid dendritic cell cytokine production
protein phosphorylation
nucleocytoplasmic transport
inflammatory response
signal transduction
neuron-neuron synaptic transmission
long-term memory
myeloid dendritic cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of protein export from nucleus
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P388650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133566 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P388650 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P388650 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P388650 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P388650 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45922 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61683 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45785 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12255 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72807 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41316 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22323 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42886 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17006 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42588 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10482 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102620 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95276 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59782 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46924 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10800816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800825 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10800814 | - | - |
Rat Genome 230 2.0 Array | 1369752_a_at | - | - |
Rat Genome 230 2.0 Array | 1369753_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369753_at | - | - |
Rat Genome 230 2.0 Array | 1369752_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369752_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369752_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369753_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369753_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800001765.rn.1 | TC1800000241.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800001764.rn.1 | TC1800000241.rn.1 | RTA TC |
Rat Expression Set 230 | 1369753_at | - | - |
Rat Expression Set 230 | 1369753_PM_at | - | - |
Rat Expression Set 230 | 1369752_a_at | - | - |
Rat Expression Set 230 | 1369752_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800814 | - | - |
Rat Genome U34 Set | M74488cds_s_at | - | - |
Rat Genome U34 Set | M63333exon_s_at | - | - |
Rat Genome U34 Set | M63333exon_s_at | - | - |
Rat Genome U34 Set | M74488cds_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20962439 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349983 | - | - |
RatRef-12 Expression BeadChip | ILMN_51541 | - | - |
