Assay Details
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012783.3 | NP_036915.1 | 1 - 2 | 159 | 74 | |
GenBank mRNA | BC059145.1 | - | 1 - 2 | 83 | 74 | |
FQ212571.1 | - | 1 - 2 | 95 | 74 | ||
FQ212582.1 | - | 1 - 2 | 103 | 74 | ||
FQ213065.1 | - | 1 - 2 | 105 | 74 | ||
FQ213626.1 | - | 1 - 2 | 95 | 74 | ||
FQ214193.1 | - | 1 - 2 | 95 | 74 | ||
FQ214232.1 | - | 1 - 2 | 100 | 74 | ||
FQ218399.1 | - | 1 - 2 | 95 | 74 | ||
FQ219910.1 | - | 1 - 2 | 79 | 74 | ||
FQ219990.1 | - | 1 - 2 | 95 | 74 | ||
FQ220920.1 | - | 1 - 2 | 103 | 74 | ||
FQ229144.1 | - | 1 - 2 | 105 | 74 | ||
FQ229755.1 | - | 1 - 2 | 92 | 74 | ||
FQ230014.1 | - | 1 - 2 | 105 | 74 | ||
FQ231850.1 | - | 1 - 2 | 105 | 74 | ||
FQ232262.1 | - | 1 - 2 | 95 | 74 | ||
FQ234420.1 | - | 1 - 2 | 105 | 74 | ||
FQ234809.1 | - | 1 - 2 | 103 | 74 | ||
FQ234829.1 | - | 1 - 2 | 95 | 74 | ||
X54640.1 | - | 286 | 74 | |||
X67215.1 | - | 1 - 2 | 80 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
