Assay Details
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012859.1 | NP_036991.1 | 1 - 2 | 1068 | 66 | |
GenBank mRNA | BC078888.1 | - | 1 - 2 | 1051 | 66 | |
U40001.1 | - | 1 - 2 | 1068 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
- Authors
- Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
- Journal
- J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
- Volume (Year)
- 201 (2009)
- Product Usage
- -
