Assay Details
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013016.2 | NP_037148.2 | 6 - 7 | 1442 | 64 | |
XM_006234951.2 | XP_006235013.1 | 5 - 6 | 1740 | 64 | ||
XM_006234952.2 | XP_006235014.1 | 6 - 7 | 2102 | 64 | ||
XM_006234953.2 | XP_006235015.1 | 6 - 7 | 2284 | 64 | ||
GenBank mRNA | AF055065.1 | - | 6 - 7 | 1306 | 64 | |
BC099773.1 | - | 6 - 7 | 1378 | 64 | ||
D38468.1 | - | 5 - 6 | 1497 | 64 | ||
D85183.1 | - | 5 - 6 | 1254 | 64 | ||
U62328.1 | - | 6 - 7 | 1442 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
