Assay Details
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017002.2 | NP_058698.2 | 1 - 2 | 203 | 76 | |
XM_006242602.2 | XP_006242664.1 | 1 - 2 | 309 | 76 | ||
GenBank mRNA | BC089810.1 | - | 1 - 2 | 203 | 76 | |
D13566.1 | - | 1 - 2 | 121 | 76 | ||
D83509.1 | - | 1 - 2 | 121 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
