Assay Details
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017312.2 | NP_059008.1 | 3 - 4 | 444 | 59 | |
GenBank mRNA | AF027954.1 | - | 2 - 3 | 354 | 59 | |
BC078871.1 | - | 3 - 4 | 444 | 59 | ||
FQ227820.1 | - | 2 - 3 | 285 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
