Assay Details
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019385.2 | NP_062258.2 | 5 - 6 | 596 | 82 | |
GenBank mRNA | AF015264.2 | - | 5 - 6 | 573 | 82 | |
BC081809.1 | - | 5 - 6 | 590 | 82 | ||
BC161850.1 | - | 5 - 6 | 596 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
