Assay Details
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145771.2 | NP_665714.2 | 6 - 7 | 527 | 107 | |
XM_008765674.1 | XP_008763896.1 | 6 - 7 | 711 | 107 | ||
GenBank mRNA | AF197128.1 | - | 6 - 7 | 521 | 107 | |
AF230096.1 | - | 5 - 6 | 529 | 107 | ||
AY738259.1 | - | 5 - 6 | 521 | 107 | ||
BC078840.1 | - | 6 - 7 | 527 | 107 | ||
FQ219187.1 | - | 5 - 6 | 516 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
