Assay Details
Gene Symbol: Gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022208.1 | NP_071544.1 | 1 - 2 | 29 | 106 | |
GenBank mRNA | AF000943.1 | - | 1 - 2 | 29 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription by RNA polymerase II
transcription initiation from RNA polymerase II promoter
biological_process
molecular_function
DNA binding
transcription coactivator activity
transcription factor binding
TBP-class protein binding
protein heterodimerization activity
RNA polymerase II-specific DNA-binding transcription factor binding
regulation of transcription, DNA-templated
transcription by RNA polymerase II
transcription initiation from RNA polymerase II promoter
biological_process
molecular_function
DNA binding
transcription coactivator activity
transcription factor binding
TBP-class protein binding
protein heterodimerization activity
RNA polymerase II-specific DNA-binding transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 468 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12161 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12161 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12161 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12161 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28285 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12524 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33064 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14201 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10891442 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891443 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891439 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891441 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891444 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891440 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10891436 | - | - |
Rat Genome 230 2.0 Array | 1369548_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369548_at | - | - |
Affymetrix HT Rat Focus Array | 1369548_at | - | - |
Affymetrix HT Rat Focus Array | 1369548_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600020839.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020844.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020845.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020846.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020842.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020841.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020848.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020843.rn.1 | TC0600002944.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020838.rn.1 | TC0600002944.rn.1 | RTA TC |
Rat Expression Set 230 | 1369548_PM_at | - | - |
Rat Expression Set 230 | 1369548_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891436 | - | - |
Rat Genome U34 Set | AF000943_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20732649 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365200 | - | - |
RatRef-12 Expression BeadChip | ILMN_68599 | - | - |
