Assay Details
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 2 - 3 | 166 | 105 | |
XM_006240996.1 | XP_006241058.1 | 2 - 3 | 268 | 105 | ||
XM_006240997.1 | XP_006241059.1 | 2 - 3 | 268 | 105 | ||
XM_006240998.2 | XP_006241060.1 | 2 - 3 | 2108 | 105 | ||
GenBank mRNA | AF368278.1 | - | 2 - 3 | 178 | 105 | |
AJ222724.1 | - | 2 - 3 | 154 | 105 | ||
BC087642.1 | - | 2 - 3 | 166 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
