Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270701.1 | NP_001257630.1 | 1 - 2 | 298 | 62 | |
NM_001270703.1 | NP_001257632.1 | 1 - 2 | 298 | 62 | ||
NM_023021.2 | NP_075410.2 | 1 - 2 | 298 | 62 | ||
XM_006228471.1 | XP_006228533.1 | 1 - 2 | 2555 | 62 | ||
GenBank mRNA | AB292803.1 | - | 1 - 2 | 156 | 62 | |
AF149250.1 | - | 1 - 2 | 156 | 62 | ||
AF156554.1 | - | 1 - 2 | 298 | 62 | ||
AF190458.1 | - | 1 - 2 | 268 | 62 | ||
AJ133438.2 | - | 1 - 2 | 156 | 62 | ||
BC128736.1 | - | 1 - 2 | 249 | 62 | ||
EU872449.1 | - | 1 - 2 | 271 | 62 | ||
EU872450.1 | - | 1 - 2 | 271 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
