Assay Details
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024154.2 | NP_077068.1 | 2 - 3 | 486 | 61 | |
XM_006257381.2 | XP_006257443.1 | 2 - 3 | 771 | 61 | ||
XM_008765794.1 | XP_008764016.1 | 2 - 3 | 771 | 61 | ||
XM_008765795.1 | XP_008764017.1 | 2 - 3 | 424 | 61 | ||
XM_008765796.1 | XP_008764018.1 | 2 - 3 | 771 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
