Assay Details
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024154.2 | NP_077068.1 | 5 - 6 | 962 | 91 | |
XM_006257378.2 | XP_006257440.1 | 3 - 4 | 1287 | 91 | ||
XM_006257381.2 | XP_006257443.1 | 5 - 6 | 1247 | 91 | ||
XM_008765793.1 | XP_008764015.1 | 3 - 4 | 1291 | 91 | ||
XM_008765796.1 | XP_008764018.1 | 5 - 6 | 1247 | 91 | ||
GenBank mRNA | AB049451.1 | - | 3 - 4 | 553 | 91 | |
AJ006519.1 | - | 3 - 4 | 1008 | 91 | ||
AJ309926.1 | - | 3 - 4 | 1012 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
