Assay Details
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030863.1 | NP_110490.1 | 2 - 3 | 193 | 93 | |
GenBank mRNA | AF004811.1 | - | 2 - 3 | 193 | 93 | |
FQ230914.1 | - | 280 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
