Assay Details
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 3 - 4 | 422 | 82 | |
XM_006246823.2 | XP_006246885.1 | 5 - 6 | 703 | 82 | ||
XM_006246824.2 | XP_006246886.1 | 4 - 5 | 598 | 82 | ||
GenBank mRNA | L06040.1 | - | 3 - 4 | 435 | 82 | |
S69383.1 | - | 3 - 4 | 422 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
