Assay Details
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031124.1 | NP_112386.1 | 8 - 9 | 749 | 96 | |
XM_008760263.1 | XP_008758485.1 | 8 - 9 | 833 | 96 | ||
XM_008760264.1 | XP_008758486.1 | 8 - 9 | 833 | 96 | ||
XM_008760265.1 | XP_008758487.1 | 8 - 9 | 833 | 96 | ||
XM_008760267.1 | XP_008758489.1 | 8 - 9 | 833 | 96 | ||
GenBank mRNA | BC168246.1 | - | 8 - 9 | 703 | 96 | |
L20820.1 | - | 8 - 9 | 749 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
