Assay Details
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031239.2 | NP_112518.1 | 1 - 2 | 181 | 83 | |
GenBank mRNA | AB008161.1 | - | 1 - 2 | 181 | 83 | |
AF009602.1 | - | 1 - 2 | 166 | 83 | ||
BC070893.1 | - | 1 - 2 | 177 | 83 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB008161.1, AF009602.1, BC070893.1, NM_031239.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
