Assay Details
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031685.2 | NP_113873.1 | 1 - 2 | 45 | 66 | |
GenBank mRNA | AF007549.1 | - | 1 - 2 | 59 | 66 | |
BC061994.1 | - | 1 - 2 | 45 | 66 | ||
U91539.1 | - | 1 - 2 | 44 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF007549.1, BC061994.1, NM_031685.2, U91539.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
